Titanium ≥99.98%, slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Leverantör: Thermo Fisher Scientific
Varning
42389.09 42389.18
42389.09EA 1750 SEK
42389.09 42389.18
Titanium ≥99.98%, slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Titanium
Purity: 99.98% CAS No.: 7440-32-6 Physical Form: ≈0.23g/slug Odour: Odourless MDL No.: MFCD00011264
Formel: Ti
Kokpunkt: 3262 °C (1013 hPa)
Smältpunkt: 1660 °C
Densitet: 4,51 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011264
CAS nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610