Titanium ≥99.99%, tråd, Ø 2.0 mm (0.08 in)
Leverantör: Thermo Scientific
Varning
14004.BU 14004.BY
14004.BUEA 2975 SEK
14004.BU 14004.BY
Titanium ≥99.99%, tråd, Ø 2.0 mm (0.08 in)
Titanium
Purity: 99.99% CAS No.: 7440-32-6 Physical Form: ≈14.2g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011264
Formel: Ti
MW: 47,88 g/mol
Kokpunkt: 3262 °C (1013 hPa)
Smältpunkt: 1660 °C
Densitet: 4,51 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011264
CAS nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610