Titanium ≥99.99%, tråd, Ø 0.25 mm (0.01 in)
Leverantör: Thermo Fisher Scientific
Varning
10392.G1 10392.G5 10392.G9
10392.G1EA 3075 SEK
10392.G1 10392.G5 10392.G9
Titanium ≥99.99%, tråd, Ø 0.25 mm (0.01 in)
Titanium
Purity: 99.99% CAS No.: 7440-32-6 Physical Form: ≈0.22g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011264
Formel: Ti
MW: 47.88 g/mol
Kokpunkt: 3262
Smältpunkt: 1660
Densitet: 4.51 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011264
CAS nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610