Titanium ≥99.98%, tråd, Ø 0.75 mm (0.03 in)
Leverantör: Thermo Fisher Scientific
Varning
00361.BU 00361.BY
00361.BUEA 1300 SEK
00361.BU 00361.BY
Titanium ≥99.98%, tråd, Ø 0.75 mm (0.03 in)
Titanium
Purity: 99.98% CAS No.: 7440-32-6 Physical Form: ≈1.99g/m, Temper: as drawn Odour: Odourless MDL No.: MFCD00011264
Formel: Ti
MW: 47.88 g/mol
Kokpunkt: 3262
Smältpunkt: 1660
Densitet: 4.51 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011264
CAS nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610