Hafnium ≥99.9%, stavar (Zr ∼ 3%), Ø 6.35 mm (0.25 in)
Leverantör: Thermo Scientific
44489.DJ 44489.DT
44489.DJEA 5375 SEK
44489.DJ 44489.DT
Hafnium ≥99.9%, stavar (Zr ∼ 3%), Ø 6.35 mm (0.25 in)
Hafnium
Purity: 99.9% CAS No.: 7440-58-6 Odour: Odourless MDL No.: MFCD00011032
Formel: Hf
MW: 178,49 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011032
CAS nummer: 7440-58-6
EINECS: 231-166-4