Hafnium ≥99.95%, tråd (Zr ∼ 3%), Ø 0.5 mm (0.02 in)
Leverantör: Thermo Fisher Scientific
42982.BU 42982.BY 42982.CC
42982.BUEA 1150 SEK
42982.BU 42982.BY 42982.CC
Hafnium ≥99.95%, tråd (Zr ∼ 3%), Ø 0.5 mm (0.02 in)
Hafnium
Purity: 99.95% CAS No.: 7440-58-6 Physical Form: ≈2.6g/m Odour: Odourless MDL No.: MFCD00011032
Formel: Hf
MW: 178.49 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011032
CAS nummer: 7440-58-6
EINECS: 231-166-4