Hafnium ≥99.9%, pieces 3-12 mm (0.12-0.47 in) (Zr ∼ 2%)
Leverantör: Thermo Scientific
39683.09 39683.18
39683.09EA 3025 SEK
39683.09 39683.18
Hafnium ≥99.9%, pieces 3-12 mm (0.12-0.47 in) (Zr ∼ 2%)
Hafnium
Purity: 99.9% CAS No.: 7440-58-6 Odour: Odourless MDL No.: MFCD00011032
Formel: Hf
MW: 178,49 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011032
CAS nummer: 7440-58-6
EINECS: 231-166-4