Yttrium ≥99.6% (REO, rare earth oxide basis), pulver -40 mesh
Leverantör: Thermo Scientific
Fara
Packaged under argon.
44147.06 44147.14
44147.06EA 3350 SEK
44147.06 44147.14
Yttrium ≥99.6% (REO, rare earth oxide basis), pulver -40 mesh
Yttrium
Purity: 99.6% CAS No.: 7440-65-5 Physical Form: Packaged under argon Odour: Odourless UN No.: 3089 MDL No.: MFCD00011468
Formel: Y
MW: 88,91 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011468
CAS nummer: 7440-65-5
EINECS: 231-174-8
UN: 3089
ADR: 4.1,II