Yttrium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.25 mm (0.01 in)
Leverantör: Thermo Scientific
13960.FF 13960.FI
13960.FFEA 3400 SEK
13960.FF 13960.FI
Yttrium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.25 mm (0.01 in)
Yttrium
Purity: 99.9% CAS No.: 7440-65-5 Physical Form: ≈0.7g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011468
Formel: Y
MW: 88,91 g/mol
Kokpunkt: 2927 °C (1013 hPa)
Smältpunkt: 1525 °C
Densitet: 4,472 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011468
CAS nummer: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240