Scandium ≥99.9%, ingot (max. 8-12% Ta)
Leverantör: Thermo Fisher Scientific
Packaged under argon.
40229.03 40229.06
40229.03EA 6250 SEK
40229.03 40229.06
Scandium ≥99.9%, ingot (max. 8-12% Ta)
Scandium
Purity: 99.9% CAS No.: 7440-20-2 Physical Form: Packaged under argon Appearance: Grey or silvery Odour: Odourless MDL No.: MFCD00016323
Formel: Sc
MW: 44.96 g/mol
Kokpunkt: 2730
Smältpunkt: 1541
Densitet: 2.989 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00016323
CAS nummer: 7440-20-2
EINECS: 231-129-2
Merck Index: 12,08537