Scandium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.25 mm (0.01 in)
Leverantör: Thermo Scientific
12424.FF 12424.FI
12424.FFEA 7850 SEK
12424.FF 12424.FI
Scandium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.25 mm (0.01 in)
Scandium
Purity: 99.9% CAS No.: 7440-20-2 Physical Form: ≈0.5g/25x25mm, Packaged in mylar bag Odour: Odourless MDL No.: MFCD00016323
Formel: Sc
MW: 44,96 g/mol
Kokpunkt: 2730 °C (1013 hPa)
Smältpunkt: 1541 °C
Densitet: 2,989 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00016323
CAS nummer: 7440-20-2
EINECS: 231-129-2
Merck Index: 12,08537