Kadmium, grovt pulver, EMSURE® ∼ 0.3-1.6 mm for analysis, suitable for filling of reductors, Supelco®
Leverantör: Merck
Fara
M-Clarity™ quality level = MQ300
1.02001.0250 1.02001.1000
1.02001.0250EA 6470 SEK
1.02001.0250 1.02001.1000
Kadmium, grovt pulver, EMSURE® ∼ 0.3-1.6 mm for analysis, suitable for filling of reductors, Supelco®
Kadmium
Formel: Cd
MW: 112.41 g/mol
Kokpunkt: 765
Smältpunkt: 321
Densitet: 8.6 g/cm3 (22 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010914
CAS nummer: 7440-43-9
EINECS: 231-152-8
UN: 3288
ADR: 6.1 III
Merck Index: 12,01649
Order Now
Specifikation testresultat
Cu (Copper) ≤ 0.002 %
Fe (Iron) ≤ 0.001 %
Pb (Lead) ≤ 0.01 %
Zn (Zinc) ≤ 0.005 %