Kadmium 99,999 Metaller basis, rods, Puratronic®, Ø 8 mm (0.3 in)
Leverantör: Thermo Fisher Scientific
Fara
43116.DM 43116.HR
43116.DMEA 3825 SEK
43116.DM 43116.HR
Kadmium 99,999 Metaller basis, rods, Puratronic®, Ø 8 mm (0.3 in)
Kadmium
Purity: 99.999% CAS No.: 7440-43-9 Physical Form: ≈0.43g/mm MDL No.: MFCD00010914
Formel: Cd
MW: 112.41 g/mol
Kokpunkt: 765
Smältpunkt: 321
Densitet: 8.642 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010914
CAS nummer: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1 II
Merck Index: 12,01649