Silver ≥99.999%, slug, Premion®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Leverantör: Thermo Fisher Scientific
42986.QJ 42986.QK
42986.QJEA 3875 SEK
42986.QJ 42986.QK
Silver ≥99.999%, slug, Premion®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Silver
Purity: 99.999% CAS No.: 7440-22-4 Physical Form: ≈2.1g/slug Odour: Characteristic MDL No.: MFCD00003397
Formel: Ag
Kokpunkt: 2212 °C (1013 hPa)
Smältpunkt: 962 °C
Densitet: 10,5 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647