Silver ≥99.99%, slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Leverantör: Thermo Fisher Scientific
42921.QI 42921.QJ 42921.QK
42921.QIEA 780 SEK
42921.QI 42921.QJ 42921.QK
Silver ≥99.99%, slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Silver
Purity: 99.99% CAS No.: 7440-22-4 Physical Form: ≈4.2g/slug Odour: Characteristic MDL No.: MFCD00003397
Formel: Ag
Kokpunkt: 2212 °C (1013 hPa)
Smältpunkt: 962 °C
Densitet: 10,5 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647