Silver ≥99.9%, spherical powder APS 0.6-2 µm
Leverantör: Thermo Fisher Scientific
41598.09 41598.18 41598.30
41598.09EA 1375 SEK
41598.09 41598.18 41598.30
Silver ≥99.9%, spherical powder APS 0.6-2 µm
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: S.A. 0.4-1.5m2/g Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397
Formel: Ag
Kokpunkt: 2212 °C (1013 hPa)
Densitet: 10,5 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III