Silver ≥99.9%, folie, annealed, Thickness: 1.0 mm (0.04 in)
Leverantör: Thermo Fisher Scientific
41452.FF 41452.FI 41452.GH
41452.FFEA 1475 SEK
41452.FF 41452.FI 41452.GH
Silver ≥99.9%, folie, annealed, Thickness: 1.0 mm (0.04 in)
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈6.6g/25x25mm, Temper: soft Odour: Characteristic MDL No.: MFCD00003397
Formel: Ag
Kokpunkt: 2212 °C (1013 hPa)
Smältpunkt: 962 °C
Densitet: 10,5 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647