Silver ≥99.9%, folie, annealed, Thickness: 0.5 mm (0.02 in)
Leverantör: Thermo Scientific
39181.FF 39181.FI 39181.GH 39181.GW
39181.FFEA 1400 SEK
39181.FF 39181.FI 39181.GH 39181.GW
Silver ≥99.9%, folie, annealed, Thickness: 0.5 mm (0.02 in)
Silver
Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈3.3g/25x25mm, Temper: soft Odour: Characteristic MDL No.: MFCD00003397
Formel: Ag
MW: 107,87 g/mol
Kokpunkt: 2212 °C (1013 hPa)
Smältpunkt: 962 °C
Densitet: 10,5 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647