Silver ≥99.9985%, tråd, Premion®, Ø 0.5 mm (0.02 in)
Leverantör: Thermo Fisher Scientific
11469.G9 11469.G1 11469.G5
11469.G9EA 17575 SEK
11469.G9 11469.G1 11469.G5
Silver ≥99.9985%, tråd, Premion®, Ø 0.5 mm (0.02 in)
Silver
Purity: 99.9985% CAS No.: 7440-22-4 Physical Form: ≈2.06g/m Odour: Characteristic MDL No.: MFCD00003397
Formel: Ag
Kokpunkt: 2212 °C (1013 hPa)
Smältpunkt: 962 °C
Densitet: 10,5 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647