Guld ≥99.995%, slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Leverantör: Thermo Fisher Scientific
40442.KG 40442.KJ
40442.KGEA 4500 SEK
40442.KG 40442.KJ
Guld ≥99.995%, slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Guld
Purity: 99.995% CAS No.: 7440-57-5 Physical Form: ≈0.49g/slug Odour: Odourless MDL No.: MFCD00003436
Electronics, vapor deposition
Formel: Au
Kokpunkt: 2800 °C (1013 hPa)
Smältpunkt: 1063 °C
Densitet: 19,3 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003436
CAS nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526