Guld ≥99.9975%, folie, Premion®, Thickness: 0.1 mm (0.004 in)
Leverantör: Thermo Scientific
11391.FF 11391.FI 11391.GH
11391.FFEA 8425 SEK
11391.FF 11391.FI 11391.GH
Guld ≥99.9975%, folie, Premion®, Thickness: 0.1 mm (0.004 in)
Guld
Purity: 99.9975+% CAS No.: 7440-57-5 Physical Form: ≈1.2g/25x25mm Odour: Odourless MDL No.: MFCD00003436
Formel: Au
MW: 196,97 g/mol
Kokpunkt: 2800 °C (1013 hPa)
Smältpunkt: 1063 °C
Densitet: 19,3 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003436
CAS nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526