Guld ≥99.9985%, folie, Premion®, Thickness: 1.0 mm (0.04 in)
Leverantör: Thermo Scientific
11388.FF 11388.FI
11388.FFEA 68900 SEK
11388.FF 11388.FI
Guld ≥99.9985%, folie, Premion®, Thickness: 1.0 mm (0.04 in)
Guld
Purity: 99.9985% CAS No.: 7440-57-5 Physical Form: ≈12g/25x25mm Odour: Odourless MDL No.: MFCD00003436
Formel: Au
MW: 196,97 g/mol
Kokpunkt: 2800 °C (1013 hPa)
Smältpunkt: 1063 °C
Densitet: 19,3 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003436
CAS nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526