Ytterbium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.1 mm (0.004 in)
Leverantör: Thermo Fisher Scientific
Fara
12437.FF 12437.FI
12437.FFEA 2500 SEK
12437.FF 12437.FI
Ytterbium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.1 mm (0.004 in)
Ytterbium
Purity: 99.9% CAS No.: 7440-64-4 Physical Form: ≈0.43g/25x25mm, Packaged in mylar under argon Odour: Odourless UN No.: 3178 MDL No.: MFCD00011286
Formel: Yb
Kokpunkt: 1427 °C (1013 hPa)
Smältpunkt: 824 °C
Densitet: 6,977 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011286
CAS nummer: 7440-64-4
EINECS: 231-173-2
UN: 3178
ADR: 4.1,III
Merck Index: 12,10239