Samarium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.25 mm (0.01 in)
Leverantör: Thermo Scientific
12420.FF 12420.FI
12420.FFEA 6500 SEK
12420.FF 12420.FI
Samarium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.25 mm (0.01 in)
Samarium
Purity: 99.9% CAS No.: 7440-19-9 Physical Form: ≈1.2g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011233
Formel: Sm
MW: 150,36 g/mol
Kokpunkt: 1900 °C (1013 hPa)
Smältpunkt: 1076,85 °C
Densitet: 7,42 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011233
CAS nummer: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498