Samarium 99.9% 20-40 mesh
Leverantör: Thermo Scientific
Fara
294780100 294780500
ACRO294780100EA 3025 SEK
ACRO294780100 ACRO294780500
Samarium 99.9% 20-40 mesh
Samarium
Formel: Sm
MW: 150,36 g/mol
Kokpunkt: 1778 °C (760 mmHg)
Smältpunkt: 1072 °C
Densitet: 7,42 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011233
CAS nummer: 7440-19-9
EINECS: 231-128-7
UN: 3132
ADR: 4.3,II
Merck Index: 12,08498
Order Now
Specifikation testresultat
Appearance Silver grey Powder or filings
Particle size 20 to 40 mesh
Total trace metal impurities ≤0.1 %