Lutetium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.1 mm (0.004 in)
Leverantör: Thermo Fisher Scientific
Fara
10228.FF 10228.FI
10228.FFEA 10300 SEK
10228.FF 10228.FI
Lutetium ≥99.9% (REO, rare earth oxide basis), folie, Thickness: 0.1 mm (0.004 in)
Lutetium
Purity: 99.9% CAS No.: 7439-94-3 Physical Form: ≈0.62g/25x25mm, Packaged in mylar under argon Appearance: Silver-grey Odour: Odourless UN No.: 3178 MDL No.: MFCD00011098
Formel: Lu
MW: 174.97 g/mol
Kokpunkt: 3000
Smältpunkt: 1651…1675 °C
Densitet: 9.849 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011098
CAS nummer: 7439-94-3
EINECS: 231-103-0
UN: 3178
ADR: 4.1 III
Merck Index: 12,05642