Cerium ≥99.8%, stavar Ø 12.7 mm (0.5 in)
Leverantör: Thermo Scientific
Fara
40284.DH 40284.DJ 40284.DM
40284.DHEA 7600 SEK
40284.DH 40284.DJ 40284.DM
Cerium ≥99.8%, stavar Ø 12.7 mm (0.5 in)
Cerium
Purity: 99.8% CAS No.: 7440-45-1 Packaged in mylar under argon Physical Form: ≈0.86g/mm, Cast rod, diameter is approximate Appearance: Silver Odour: Odourless UN No.: 1333 MDL No.: MFCD00010924
Formel: Ce
MW: 140,12 g/mol
Kokpunkt: 3468 °C (1013 hPa)
Smältpunkt: 795 °C
Densitet: ∼6,8 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010924
CAS nummer: 7440-45-1
EINECS: 231-154-9
UN: 1333
ADR: 4.1,II