Vismut ≥99.999%, spherical shots 1-5 mm (0.04-0.20 in)
Leverantör: Thermo Fisher Scientific
Ampuled under argon
38619.09 38619.18
38619.09EA 4300 SEK
38619.09 38619.18
Vismut ≥99.999%, spherical shots 1-5 mm (0.04-0.20 in)
Vismut
Purity: 99.999% CAS No.: 7440-69-9 Physical Form: Ampouled under argon Odour: Odourless MDL No.: MFCD00134033
Formel: Bi
MW: 208.98 g/mol
Kokpunkt: 1490
Smältpunkt: 271…272 °C
Densitet: 9.78 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134033
CAS nummer: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297