Antimon ≥99.8%, broken rod, Length: various, Ø 12.7 mm (0.50 in)
Leverantör: Thermo Scientific
Varning
45047.A3 45047.36
45047.A3EA 16425 SEK
45047.A3 45047.36
Antimon ≥99.8%, broken rod, Length: various, Ø 12.7 mm (0.50 in)
Antimony
Purity: 99.8% CAS No.: 7440-36-0 Physical Form: Broken pieces from cast rod Odour: Odourless MDL No.: MFCD00134030
Formel: Sb
MW: 121,76 g/mol
Kokpunkt: 1637 °C (1013 hPa)
Smältpunkt: 630 °C
Densitet: 6,684 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134030
CAS nummer: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III
Merck Index: 12,00733