Tenn ≥99.999%, slug, Puratronic®, Ø 11 mm (0.43 in), Thickness: 4 mm (0.2 in)
Leverantör: Thermo Fisher Scientific
14134.KD 14134.KK
14134.KDEA 15725 SEK
14134.KD 14134.KK
Tenn ≥99.999%, slug, Puratronic®, Ø 11 mm (0.43 in), Thickness: 4 mm (0.2 in)
Tenn
Purity: 99.999% CAS No.: 7440-31-5 Physical Form: ≈2.6g Odour: Odourless MDL No.: MFCD00133862
Formel: Sn
MW: 118.71 g/mol
Kokpunkt: 2270
Smältpunkt: 232
Densitet: 7.31 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00133862
CAS nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587