Tenn ≥99.998%, tråd, Puratronic®, Ø 1.0 mm (0.04 in)
Leverantör: Thermo Fisher Scientific
11905.BW 11905.CB 11905.CD
11905.BWEA 1075 SEK
11905.BW 11905.CB 11905.CD
Tenn ≥99.998%, tråd, Puratronic®, Ø 1.0 mm (0.04 in)
Tenn
Purity: 99.998% CAS No.: 7440-31-5 Physical Form: ≈5.72g/m Odour: Odourless MDL No.: MFCD00133862
Formel: Sn
MW: 118.71 g/mol
Kokpunkt: 2270
Smältpunkt: 232
Densitet: 7.31 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00133862
CAS nummer: 7440-31-5
EINECS: 23-141-8
Merck Index: 12,09587