Tenn ≥99.9985%, folie, Puratronic®, Thickness: 1.0 mm (0.04 in)
Leverantör: Thermo Fisher Scientific
11457.FG 11457.FY 11457.HL
11457.FGEA 2025 SEK
11457.FG 11457.FY 11457.HL
Tenn ≥99.9985%, folie, Puratronic®, Thickness: 1.0 mm (0.04 in)
Tenn
Purity: 99.9985% CAS No.: 7440-31-5 Physical Form: ≈9.1g/25x50mm Odour: Odourless MDL No.: MFCD00133862
Formel: Sn
MW: 118.71 g/mol
Kokpunkt: 2270
Smältpunkt: 232
Densitet: 7.31 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00133862
CAS nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587