Tenn 99.8%, pulver -325 mesh
Leverantör: Thermo Fisher Scientific
Varning
317801000 317805000
ACRO317801000EA 825 SEK
ACRO317801000 ACRO317805000
Tenn 99.8%, pulver -325 mesh
Tenn
Formel: Sn
Kokpunkt: 2270 °C (760 mmHg)
Densitet: 7,3 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00133862
CAS nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587
Order Now
Specifikation testresultat
Appearance Grey Powder
Total trace metal impurities ≤0.2 %