Grafit, plate, highly oriented pyrolytic graphite (HOPG) 10x10x(1.6 min) mm (0.394x0.394x0.079 in)
Leverantör: Thermo Fisher Scientific
43835.KF 43835.KJ
43835.KFEA 14300 SEK
43835.KF 43835.KJ
Grafit, plate, highly oriented pyrolytic graphite (HOPG) 10x10x(1.6 min) mm (0.394x0.394x0.079 in)
Grafit
Physical Form: 0.8 +/-0.2° mosaic angle MDL No.: MFCD00144065
Formel: C
Smältpunkt: 3827 °C
Densitet: 2,09…2.23 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00144065
CAS nummer: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560