Grafit, rods, pyrolytic coated, Length: 152 mm (6 in), Ø 2 mm (0.08 in)
Leverantör: Thermo Fisher Scientific
43670.KJ 43670.KG
43670.KJEA 13300 SEK
43670.KJ 43670.KG
Grafit, rods, pyrolytic coated, Length: 152 mm (6 in), Ø 2 mm (0.08 in)
Grafit
Physical Form: ≈0.8g/pc MDL No.: MFCD00144065
Formel: C
MW: 12.01 g/mol
Smältpunkt: 3827
Densitet: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00144065
CAS nummer: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560