Aktivt kol 2mm & down
Leverantör: Thermo Scientific
Synonyms: Carbon activated, Activated carbon, Activated charcoal
10926.22 10926.30
10926.22EA 995 SEK
10926.22 10926.30
Aktivt kol 2mm & down
Aktivt kol
MDL No.: MFCD00133992
For liquid purification
Formel: C
MW: 12,01 g/mol
Smältpunkt: 3550 °C
Densitet: 2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00133992
CAS nummer: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818