Aktivt kol, NORIT® SA 2, decolorizing
Leverantör: Thermo Scientific
Synonyms: Carbon activated, Activated carbon, Activated charcoal
404035000 404030025
ACRO404035000EA 970 SEK
ACRO404035000 ACRO404030025
Aktivt kol, NORIT® SA 2, decolorizing
Aktivt kol
Formel: C
MW: 12,01 g/mol
Smältpunkt: 3652 °C
Densitet: 2 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00133992
CAS nummer: 7440-44-0
EINECS: 231-153-3
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818
Order Now
Specifikation testresultat
Appearance Black Powder
Loss on drying ≤10 %
Molasses number ≤390
GC ≥95 %