Carbon ≥99.96%, yarn, woven from 0.076 mm (0.003 in) dia fibers
Leverantör: Thermo Fisher Scientific
39721.G5 39721.G9
39721.G5EA 890 SEK
39721.G5 39721.G9
Carbon ≥99.96%, yarn, woven from 0.076 mm (0.003 in) dia fibers
Carbon
MDL No.: MFCD00133992
Formel: C
Smältpunkt: 3727 °C
Densitet: 3,65 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00133992
CAS nummer: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818