Indium ≥99.9%, pulver -100 mesh
Leverantör: Thermo Fisher Scientific
Varning
45122.14 45122.06
45122.14EA 10200 SEK
45122.14 45122.06
Indium ≥99.9%, pulver -100 mesh
Indium
Purity: 99.9% CAS No.: 7440-74-6 Molecular Formula: In Formula Weight: 114.82 Physical Form: Powder Density: 7.31 Appearance: Silver-grey Odour: Odourless Melting Point: 156.6° Boiling Point: 2072° UN No.: 3089 MDL No.: MFCD00134048
Formel: In
MW: 114.82 g/mol
Kokpunkt: 2072 °C
Smältpunkt: 156.6 °C
Densitet: 7.31
Storage Temperature: Omgivande
MDL Number: MFCD00134048
CAS nummer: 7440-74-6
EINECS: 231-180-0
UN: 3089
ADR: 4.1 III
Merck Index: 13,04971