Indium ≥99.998%, slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Leverantör: Thermo Fisher Scientific
43414.22 43414.14
43414.22EA 14725 SEK
43414.22 43414.14
Indium ≥99.998%, slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Indium
Purity: 99.998% CAS No.: 7440-74-6 Formula Weight: 114.818 Physical Form: ≈1.5g/slug Density: 7.310G/CC Odour: Odourless Boiling Point: 2072 deg C MDL No.: MFCD00134048
Formel: In
Kokpunkt: 2072 °C
Densitet: 7,310G/CC
Storage Temperature: Omgivande
MDL Number: MFCD00134048
CAS nummer: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971