Indium ≥99.998%, tråd, Puratronic®, Ø 1.0 mm (0.04 in)
Leverantör: Thermo Scientific
11462.G1 11462.G5 11462.G9
11462.G1EA 3425 SEK
11462.G1 11462.G5 11462.G9
Indium ≥99.998%, tråd, Puratronic®, Ø 1.0 mm (0.04 in)
Indium
Purity: 99.998+% CAS No.: 7440-74-6 Physical Form: ≈5.74g/m Odour: Odourless MDL No.: MFCD00134048
Formel: In
MW: 114,82 g/mol
Kokpunkt: 2000 °C (1013 hPa)
Smältpunkt: 156 °C
Densitet: 7,3 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134048
CAS nummer: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971