Indium ≥99.9975%, folie, Puratronic®, Thickness: 0.25 mm (0.01 in)
Leverantör: Thermo Fisher Scientific
11385.FI 11385.GH
11385.FIEA 3950 SEK
11385.FI 11385.GH
Indium ≥99.9975%, folie, Puratronic®, Thickness: 0.25 mm (0.01 in)
Indium
Purity: 99.9975% CAS No.: 7440-74-6 Physical Form: ≈4.6g/50x50mm Odour: Odourless MDL No.: MFCD00134048
Formel: In
MW: 114.82 g/mol
Kokpunkt: 2000
Smältpunkt: 156
Densitet: 7.3 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134048
CAS nummer: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971