Gallium ≥99.999%, ingot
Leverantör: Thermo Fisher Scientific
Varning
10187.18 10187.30 10187.36 10187.14
10187.18EA 10525 SEK
10187.18 10187.30 10187.36 10187.14
Gallium ≥99.999%, ingot
Gallium
Purity: 99.999% CAS No.: 7440-55-3 Molecular Formula: Requires overnight shipping with cold pack Storage Temperature: 2°C to 8°C Appearance: Silver grey MDL No.: MFCD00134045
Formel: Ga
MW: 69.72 g/mol
Kokpunkt: 2403
Smältpunkt: 29
Densitet: 5.904 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00134045
CAS nummer: 7440-55-3
EINECS: 231-163-8
UN: 2803
ADR: 8 III
Merck Index: 12,04364