Aluminium ≥99.9999%, folie, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Aluminum
44330.GH 44330.HP
44330.GHEA 3800 SEK
44330.GH 44330.HP
Aluminium ≥99.9999%, folie, Puratronic®, annealed, Thickness: 0.2 mm (0.008 in)
Aluminium
Purity: 99.9999% CAS No.: 7429-90-5 Physical Form: ≈5.4g/100x100mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Kokpunkt: 2327 °C (1013 hPa)
Smältpunkt: 660 °C
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321