Aluminium ≥99.9998%, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Aluminum
44327.09 44327.18
44327.09EA 2550 SEK
44327.09 44327.18
Aluminium ≥99.9998%, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Aluminium
Purity: 99.9998% CAS No.: 7429-90-5 Physical Form: ≈0.14g/slug Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Kokpunkt: 2327 °C (1013 hPa)
Smältpunkt: 660 °C
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321