Aluminium ≥99.9995%, tråd, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Aluminum
43427.H4 43427.G9 43427.G5
43427.H4EA 29250 SEK
43427.H4 43427.G9 43427.G5
Aluminium ≥99.9995%, tråd, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Aluminium
Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈2.1g/m, Temper: soft Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Kokpunkt: 2327 °C (1013 hPa)
Smältpunkt: 660 °C
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321