Aluminium ≥99.9995%, folie, Puratronic®, annealed, Thickness: 0.25 mm (0.01 in)
Leverantör: Thermo Scientific
Synonyms: Aluminum
43424.GH 43424.HP 43424.FI
43424.GHEA 3725 SEK
43424.GH 43424.HP 43424.FI
Aluminium ≥99.9995%, folie, Puratronic®, annealed, Thickness: 0.25 mm (0.01 in)
Aluminium
Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈1.7g/50x50mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029
Formel: Al
MW: 26,98 g/mol
Kokpunkt: 2327 °C (1013 hPa)
Smältpunkt: 660 °C
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321