Aluminium ≥99.9995%, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Aluminum
43357.22 43357.14
43357.22EA 6875 SEK
43357.22 43357.14
Aluminium ≥99.9995%, slug, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Aluminium
Purity: 99.9995% CAS No.: 7429-90-5 Physical Form: ≈0.54g/slug Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Kokpunkt: 2327 °C (1013 hPa)
Smältpunkt: 660 °C
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321