Aluminium ≥99.99%, slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Aluminum
42258.18 42258.30
42258.18EA 3875 SEK
42258.18 42258.30
Aluminium ≥99.99%, slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Aluminium
Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈0.14g/slug Odour: Odourless MDL No.: MFCD00134029
Formel: Al
MW: 26.98 g/mol
Kokpunkt: 2327
Smältpunkt: 660
Densitet: 2.7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321