Aluminium ≥99.999%, sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Leverantör: Thermo Scientific
Synonyms: Aluminum
40827.KS 40827.KU
40827.KSEA 12450 SEK
40827.KS 40827.KU
Aluminium ≥99.999%, sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Aluminium
Purity: 99.999% CAS No.: 7429-90-5 Odour: Odourless MDL No.: MFCD00134029
Formel: Al
MW: 26,98 g/mol
Kokpunkt: 2327 °C (1013 hPa)
Smältpunkt: 660 °C
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321